Learn More
Biomatik Corporation Human FBLN5 (Fibulin-5) ELISA Kit
Supplier:  Biomatik Corporation EKF57226
Encompass_Preferred
Human FBLN5 (Fibulin-5) ELISA Kit for detecting FBLN5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9UBX5. Target Synonyms: FBLN5; Fibulin-5; DANCE; EVEC; UP50; ARMD3; FIBL-5; ; fibulin 5; Urine p50 protein; Developmental arteries and neural crest EGF-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7892