Learn More
Biomatik Corporation Human SHBG (Sex hormone-binding globulin) ELISA Kit
Supplier:  Biomatik Corporation EKF57252
Encompass_Preferred
Human SHBG (Sex hormone-binding globulin) ELISA Kit for detecting SHBG in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pmol/l. Sensitivity: 9.375pmol/l. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04278. Target Synonyms: SHBG; ABP; androgen binding protein; SBP; sex hormone-binding globulin; Sex steroid-binding protein; TeBG; TEBG; Testis-specific androgen-binding protein; testosterone-binding beta-globulin; Testosterone-estradiol-binding globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7900