Learn More
Biomatik Corporation Human PODXL (Podocalyxin) ELISA Kit
Supplier:  Biomatik Corporation EKF57282
Encompass_Preferred
Human PODXL (Podocalyxin) ELISA Kit for detecting PODXL in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O00592. Target Synonyms: PCX; Podocalyxin; PODXL; PCLP; PCLP1; Podocalyxin-like protein 1; PC; PCLP-1; GCTM-2 antigen; Gp200. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7904