Learn More
Biomatik Corporation Human PYGB (Glycogen phosphorylase, brain form) ELISA Kit
Supplier:  Biomatik Corporation EKF57296
Encompass_Preferred
Human PYGB (Glycogen phosphorylase, brain form) ELISA Kit for detecting PYGB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P11216. Target Synonyms: PYGB; Glycogen phosphorylase brain form; Glycogen Phosphorylase BB; GPBB; Brain glycogen phosphorylase; Glycogen phosphorylase B; Glycogen Phosphorylase Isoenzyme BB; Phosphorylase glycogen brain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7914