Learn More
Biomatik Corporation Human PARP1 (Poly [ADP-ribose] polymerase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57300
Encompass_Preferred
Human PARP1 (Poly [ADP-ribose] polymerase 1) ELISA Kit for detecting PARP1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P09874. Target Synonyms: PARP1; ADPRT; PPOL; Poly [ADP-ribose] polymerase 1; PARP-1; Poly[ADP-ribose] synthase 1; ADPRT 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7916