Learn More
Biomatik Corporation Human BECN1 (Beclin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF57309
Encompass_Preferred
Human BECN1 (Beclin-1) ELISA Kit for detecting BECN1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q14457. Target Synonyms: BECN1; Beclin 1; ATG6; GT197; VPS30; ATG6 autophagy related 6 homolog; beclin 1; autophagy related; beclin-1; Coiled-coil myosin-like BCL2-interacting protein; Protein GT197. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7918