Learn More
Biomatik Corporation Human PTN (Pleiotrophin) ELISA Kit
Supplier:  Biomatik Corporation EKF57342
Encompass_Preferred
Human PTN (Pleiotrophin) ELISA Kit for detecting PTN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21246. Target Synonyms: PTN; Pleiotrophin; Osteoblast-specific factor 1; OSF-1; Heparin-binding neurite outgrowth-promoting factor 1; HBNF-1; Heparin-binding growth-associated molecule; HB-GAM; Heparin-binding brain mitogen; HBBM; Heparin-binding growth factor 8; HBGF-8; neurite growth-promoting factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7923