Learn More
Biomatik Corporation Human IDO1 (Indoleamine 2,3-dioxygenase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57381
Encompass_Preferred
Human IDO1 (Indoleamine 2,3-dioxygenase 1) ELISA Kit for detecting IDO1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P14902. Target Synonyms: IDO1; IDO; INDO; IDO-1; Indoleamine-pyrrole 2; 3-dioxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7932