Learn More
Biomatik Corporation Human MAPT (Microtubule-associated protein tau) ELISA Kit
Supplier:  Biomatik Corporation EKF57422
Encompass_Preferred
Human MAPT (Microtubule-associated protein tau) ELISA Kit for detecting MAPT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10636. Target Synonyms: MAPT; Microtubule-associated protein tau; TAU; MSTD; PPND; DDPAC; MAPTL; MTBT1; MTBT2; FTDP-17; Neurofibrillary tangle protein; Paired helical filament-tau. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7940