Learn More
Biomatik Corporation Human DNMT1 (DNA (cytosine-5)-methyltransferase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57443
Encompass_Preferred
Human DNMT1 (DNA (cytosine-5)-methyltransferase 1) ELISA Kit for detecting DNMT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P26358. Target Synonyms: DNMT1; DNA; cytosine-5-methyltransferase 1; CXXC9; MCMT; AIM; CXXC9CXXC finger protein 9; CXXC-type zinc finger protein 9; DNA; cytosine-5--methyltransferase 1; DNA; cytosine-5-methyltransferase 1; DNA methyltransferase 1; DNA methyltransferase HsaI; DNA MTase HsaI; DNMT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7943