Learn More
Biomatik Corporation Human COL4 (Collagen Type IV) ELISA Kit
Supplier:  Biomatik Corporation EKF57575
Encompass_Preferred
Human COL4 (Collagen Type IV) ELISA Kit for detecting COL4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02462; P08572; Q01955; P53420; P29400; Q14031. Target Synonyms: COL4; Collagen Type IV. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7972