Learn More
Biomatik Corporation Human GA (Glycated Albumin) ELISA Kit
Supplier:  Biomatik Corporation EKF57615
Encompass_Preferred
Human GA (Glycated Albumin) ELISA Kit for detecting GA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100pmol/ml. Sensitivity: 0.938pmol/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02768. Target Synonyms: GA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7979