Learn More
Biomatik Corporation Human PPAR-γ (Peroxisome ProlifeRator Activated Receptor Gamma) ELISA Kit
Supplier:  Biomatik Corporation EKF57696
Encompass_Preferred
Human PPAR-γ (Peroxisome ProlifeRator Activated Receptor Gamma) ELISA Kit for detecting PPAR-γ in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P37231. Target Synonyms: PPAR-γ; PPARG; NR1C3; Peroxisome prolifeRator-activated receptor gamma; CIMT1; Nuclear receptor subfamily 1 group C member 3; peroxisome prolifeRator-activated receptor gamma 1; PPAR gamma; PPARG1; peroxisome prolifeRative activated receptor; gamma; PPARG2; PPARgamma; PPAR-gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7989