Learn More
Biomatik Corporation Human SPA (Pulmonary Surfactant Associated Protein A) ELISA Kit
Supplier:  Biomatik Corporation EKF57719
Encompass_Preferred
Human SPA (Pulmonary Surfactant Associated Protein A) ELISA Kit for detecting SPA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8IWL2. Target Synonyms: SPA; Pulmonary Surfactant Associated Protein A; SFTPA1; COLEC4; PSAP; SFTP1; SFTPA; SFTPA1B; PSP-A; PSPA; SP-A; SP-A1; Collectin-4; 35 kDa pulmonary surfactant-associated protein; Alveolar proteinosis protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7994