Learn More
Biomatik Corporation Human MAP1LC3B (Microtubule-associated proteins 1A/1B light chain 3B) ELISA Kit
Supplier:  Biomatik Corporation EKF58014
Encompass_Preferred
Human MAP1LC3B (Microtubule-associated proteins 1A/1B light chain 3B) ELISA Kit for detecting MAP1LC3B in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9GZQ8. Target Synonyms: MAP1LC3B; Microtubule-associated protein 1 light chain 3 beta; MAP1A; MAP1B light chain 3 B; MAP1A; MAP1B LC3 B; MAP1 light chain 3-like protein 2; Autophagy-related protein LC3 B; Autophagy-related ubiquitin-like modifier LC3 B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8008