Learn More
Biomatik Corporation PD1 (Protectin D1) ELISA Kit
Supplier:  Biomatik Corporation EKF58060
Encompass_Preferred
PD1 (Protectin D1) ELISA Kit for detecting PD1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: PD1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8030