Learn More
Biomatik Corporation Mouse NT-proBNP (N-terminal pro-Brain Natriuretic Peptide) ELISA Kit
Supplier:  Biomatik Corporation EKF58162
Encompass_Preferred
Mouse NT-proBNP (N-terminal pro-Brain Natriuretic Peptide) ELISA Kit for detecting NT-proBNP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P40753. Target Synonyms: NT-proBNP; N-Terminal Pro-Brain Natriuretic Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8050