Learn More
Biomatik Corporation Pig Cortisol ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC31445
Encompass_Preferred
Pig Cortisol Elisa Kit for detecting Cortisol in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.049-200ng/ml. Sensitivity: 0.049ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-7462