Learn More
Biomatik Corporation Human STAT3 (Signal transducer and activator of transcription 3) ELISA Kit
Supplier:  Biomatik Corporation EKF58198
Encompass_Preferred
Human STAT3 (Signal transducer and activator of transcription 3) ELISA Kit for detecting STAT3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P40763. Target Synonyms: STAT3; Signal Transducer and Activator of Transcription 3; Acute-phase response factor; APRFMGC16063; DNA-binding protein APRF; HIES. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8062