Learn More
Biomatik Corporation Human DLL4 (Delta-like protein 4) ELISA Kit
Supplier:  Biomatik Corporation EKF58286
Encompass_Preferred
Human DLL4 (Delta-like protein 4) ELISA Kit for detecting DLL4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9NR61. Target Synonyms: Delta-like protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8087