Learn More
Biomatik Corporation Rat GH (Growth Hormone) ELISA Kit
Supplier:  Biomatik Corporation EKF58315
Encompass_Preferred
Rat GH (Growth Hormone) ELISA Kit for detecting GH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 9.375-600pg/ml. Sensitivity: 5.625pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01244. Target Synonyms: GH; Growth Hormone; GH1; GH-N; GHN; hGH-N; Somatotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8095