Learn More
Biomatik Corporation Rat PGE2 (Prostaglandin E2) ELISA Kit
Supplier:  Biomatik Corporation EKF58374
Encompass_Preferred
Rat PGE2 (Prostaglandin E2) ELISA Kit for detecting PGE2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: PGE2; Prostaglandin E2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8108