Learn More
Biomatik Corporation Human MT-RNR2 Like Protein 1 (MTRNR2L1) ELISA Kit
Supplier:  Biomatik Corporation EKU11298
Encompass_Preferred
Human MT-RNR2 Like Protein 1 (MTRNR2L1) ELISA Kit for detecting MTRNR2L1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P0CJ68. Target Synonyms: HN1; Humanin Like Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6886