Learn More
Biomatik Corporation Vitamin B2 (VB2) ELISA Kit
Supplier:  Biomatik Corporation EKU11524
Encompass_Preferred
Vitamin B2 (VB2) ELISA Kit for detecting VB2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 7.41-600ng/ml. Sensitivity: 2.61ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Riboflavin; Riboflavine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6941