Learn More
Biomatik Corporation Penicillin (PC) ELISA Kit
Supplier:  Biomatik Corporation EKU11586
Encompass_Preferred
Penicillin (PC) ELISA Kit for detecting PC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 12.35-1,000ng/ml. Sensitivity: 5.23ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6960