Learn More
Biomatik Corporation Horse Collagen Type III (COL3) ELISA Kit
Supplier:  Biomatik Corporation EKU11662
Encompass_Preferred
Horse Collagen Type III (COL3) ELISA Kit for detecting COL3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.60ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6980