Learn More
Biomatik Corporation Human Transglutaminase 2 (TGM2) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11975
Encompass_Preferred
Human Transglutaminase 2 (TGM2) Mini sample ELISA Kit for detecting TGM2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.241ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21980. Gene ID: 7052. Target Synonyms: tTG; TG2; C Polypeptide,Protein-Glutamine-Gamma-Glutamyltransferase; Transglutaminase 2, Tissue; Transglutaminase C; Transglutaminase H. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7042