Learn More
Biomatik Corporation Mouse Oxoglutarate Dehydrogenase (OGDH) ELISA Kit
Supplier:  Biomatik Corporation EKU12051
Encompass_Preferred
Mouse Oxoglutarate Dehydrogenase (OGDH) ELISA Kit for detecting OGDH in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.60ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60597. Gene ID: 18293. Target Synonyms: AKGDH; E1k; OGDC; Alpha-Ketoglutarate Dehydrogenase,Lipoamide; 2-Oxoglutarate Dehydrogenase E1 Component,Mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7059