Learn More
Biomatik Corporation Human Cytokeratin 20, Ck20 Elisa Kit
Supplier:  Biomatik Corporation EKC33392
Encompass_Preferred
Human Cytokeratin 20, Ck20 Elisa Kit for detecting Keratin 20 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35900; KRT20. Target Synonyms: CD20; CK 20; CK-20; CK20; Cytokeratin-20; Cytokeratin20; K1C20_HUMAN; K20; KA20; Keratin 20; keratin 20; type I; keratin 21; rat; homolog of; Keratin; Keratin type I cytoskeletal 20; Keratin-20; Keratin20; KRT 20; KRT 21; KRT20; KRT21; MGC35423; OTTHUMP00000164518; Protein IT; type I cytoskeletal 20. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7394