Learn More
Biomatik Corporation Human D-Lactate Dehydrogenase, D-Ldh Elisa Kit
Supplier:  Biomatik Corporation EKC33479
Encompass_Preferred
Human D-Lactate Dehydrogenase, D-Ldh Elisa Kit for detecting Lactate Dehydrogenase D in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000mIU/ml. Sensitivity: 7.8mIU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q86WU2; LDHD. Target Synonyms: D lactate dehydrogenase; DLD; Lactate dehydrogenase D; Ldhd; LDHD_HUMAN; MGC57726; mitochondrial; Probable D lactate dehydrogenase mitochondrial; Probable D-lactate dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7399