Learn More
Biomatik Corporation Human Erythropoietin, Epo Elisa Kit
Supplier:  Biomatik Corporation EKC33631
Encompass_Preferred
Human Erythropoietin, Epo Elisa Kit for detecting Erythropoietin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500mIU/ml. Sensitivity: 1.95mIU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01588; EPO. Target Synonyms: EP; MGC138142; MVCD2; epoetin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7407