Learn More
Biomatik Corporation Human Estradiol Receptor, Er Elisa Kit
Supplier:  Biomatik Corporation EKC33633
Encompass_Preferred
Human Estradiol Receptor, Er Elisa Kit for detecting Estrogen Receptor 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P03372; ESR1. Target Synonyms: 7*/654 isoform; 7*/819 2 isoform; 7*/822 isoform; 8*/901 isoform; 8*/941 isoform; DKFZp686N23123; ER alpha; ER; ER-alpha; Era; ESR; ESR1; ESR1_HUMAN; ESRA; Estradiol receptor; Estrogen nuclear receptor alpha; Estrogen receptor 1; Estrogen receptor alpha 3*; 4; 5; 6; 7*/822 isoform; Estrogen receptor alpha; Estrogen receptor alpha delta 3*; 4; 5; 6; 7*; 8*/941 isoform; Estrogen receptor alpha delta 3*; 4; 5; 6; 7*/819 2 isoform; Estrogen receptor alpha delta 4 +49 isoform; Estrogen receptor alpha delta 4*; 5; 6; 7*/654 isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7409