Learn More
Biomatik Corporation Human Ferritin, Heavy Polypeptide (Fth) Elisa Kit
Supplier:  Biomatik Corporation EKC33672
Encompass_Preferred
Human Ferritin, Heavy Polypeptide (Fth) Elisa Kit for detecting Ferritin, Heavy Polypeptide 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.45-30ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02794; FTH1. Target Synonyms: Apoferritin; Cell proliferation inducing gene 15 protein; Cell proliferation-inducing gene 15 protein; F HC; Ferritin H subunit; Ferritin heavy chain; Ferritin heavy polypeptide 1; FHC; FRIH; FRIH_HUMAN; FTH 1; FTH; FTH1; FTH1 protein; FTHL 6; FTHL6; Iron overload autosomal dominant; MGC104426; N-terminally processed; OK/SW-cl.84; PIG 15; PIG15; Placenta immunoregulatory factor; PLIF; Proliferation inducing gene 15 protein; Proliferation inducing protein 15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7411