Learn More
Biomatik Corporation Human Glucagon-Like Peptide-1, Glp-1 Elisa Kit
Supplier:  Biomatik Corporation EKC33823
Encompass_Preferred
Human Glucagon-Like Peptide-1, Glp-1 Elisa Kit for detecting Glucagon-Like Peptide-1, Glp-1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 2.3-40ng/ml. Sensitivity: 1.4ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7423