Learn More
Biomatik Corporation Human Glutathione Peroxidase 3 (Gpx3) Elisa Kit
Supplier:  Biomatik Corporation EKC33854
Encompass_Preferred
Human Glutathione Peroxidase 3 (Gpx3) Elisa Kit for detecting Glutathione Peroxidase 3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000uIU/ml. Sensitivity: 3.9uIU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P22352; GPX3. Target Synonyms: Extracellular glutathione peroxidase; Glutathione peroxidase 3 (plasma); Glutathione peroxidase 3; GPX 3; GPx P; GPx-3; GPx-P; GPX3; GPX3_HUMAN; GPXP; GSHPx 3; GSHPx P; GSHPx-3; GSHPx-P; Plasma glutathione peroxidase 3; Plasma glutathione peroxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7426