Learn More
Biomatik Corporation Human Granzyme B (Gzmb) Elisa Kit
Supplier:  Biomatik Corporation EKC33903
Encompass_Preferred
Human Granzyme B (Gzmb) Elisa Kit for detecting Granzyme B (Granzyme 2, Cytotoxic T-Lymphocyte-Associated Serine Esterase 1) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10144; GZMB. Target Synonyms: CCPI; CGL-1; CGL1; CSP-B; CSPB; CTLA1; CTSGL1; HLP; SECT; T-cell serine protease 1-3E; cathepsin G-like 1; cytotoxic serine protease B; fragmentin 2; granzyme B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7428