Learn More
Biomatik Corporation Human Heat Shock Protein 72 (Hsp-72) Elisa Kit
Supplier:  Biomatik Corporation EKC33952
Encompass_Preferred
Human Heat Shock Protein 72 (Hsp-72) Elisa Kit for detecting Heat Shock Protein 72(Hsp-72) in serum, plasma, cell culture supernates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P0DMV8; HSPA1A. Target Synonyms: HSPA1A; HSP72; HSPA1; HSX70Heat shock 70 kDa protein 1A; Heat shock 70 kDa protein 1; HSP70-1; HSP70.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7430