Learn More
Biomatik Corporation Human Homocysteine(HCY)ELISA Kit
Supplier:  Biomatik Corporation EKC34070
Encompass_Preferred
Sample Type: serum, plasma, urine, tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78 nmol/mL-50 nmol/mL. Sensitivity: 0.195 nmol/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7435