Learn More
Biomatik Corporation Human Inhibin B, Inh-B Elisa Kit
Supplier:  Biomatik Corporation EKC34128
Encompass_Preferred
Human Inhibin B, Inh-B Elisa Kit for detecting Inhibin B, Inh-B in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 4-800pg/ml. Sensitivity: 2pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7442