Learn More
Biomatik Corporation Human Insulin-Like Growth Factors 2, Igf-2 Elisa Kit
Supplier:  Biomatik Corporation EKC34155
Encompass_Preferred
Human Insulin-Like Growth Factors 2, Igf-2 Elisa Kit for detecting Insulin-Like Growth Factor 2 (Somatomedin A) in serum, plasma, cell culture supernates, urine, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01344; IGF2. Target Synonyms: C11orf43; IGF 2; IGF II; IGF-II; IGF2; IGF2_HUMAN; IGFII; INSIGF; insulin like growth factor 2 (somatomedin A); Insulin like Growth Factor 2; Insulin like growth factor II; Insulin like growth factor II precursor; Insulin like growth factor type 2; pp9974; Preptin; putative insulin like growth factor II associated protein; Somatomedin A; Somatomedin-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7446