Learn More
Biomatik Corporation Human Integrin Αm, Itg/Cd11B Elisa Kit
Supplier:  Biomatik Corporation EKC34176
Encompass_Preferred
Human Integrin Αm, Itg/Cd11B Elisa Kit for detecting Integrin, Alpha M (Complement Component 3 Receptor 3 Subunit) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P11215; ITGAM. Target Synonyms: antigen CD11b (p170); Antigen CD11b p170; CD11 antigen like family member B; CD11 antigen-like family member B; CD11b; CD11b/CD18; CD49d; Cell surface glycoprotein MAC-1 subunit alpha; Complement component 3 receptor 3 subunit; Complement Component Receptor 3 Alpha; Complement receptor type 3; Complement receptor type 3; alpha subunit; CR 3 alpha chain (CR3A); CR 3 alpha chain; CR-3 alpha chain; CR3; CR3A; F730045J24Rik; Integrin Alpha M; Integrin alpha M chain; Integrin alpha-M; Integrin beta 2 alpha subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7449