Learn More
Biomatik Corporation Human Interleukin 15, Il-15 Elisa Kit
Supplier:  Biomatik Corporation EKC34225
Encompass_Preferred
Human Interleukin 15, Il-15 Elisa Kit for detecting Interleukin 15 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P40933; IL15. Target Synonyms: IL 15; IL-15; IL15; IL15_HUMAN; Interleukin 15; Interleukin-15; Interleukin15; MGC9721. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7451