Learn More
Biomatik Corporation Human Interleukin 27, Il-27 Elisa Kit
Supplier:  Biomatik Corporation EKC34240
Encompass_Preferred
Human Interleukin 27, Il-27 Elisa Kit for detecting Interleukin 27 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8NEV9; IL27. Target Synonyms: EBI3; IL 27; IL 27 p28 subunit; IL 27A; IL 30; IL-27 subunit alpha; IL-27-A; IL27; IL27-A; IL27A; IL27A_HUMAN; IL27p28; IL30; Interleukin 27; Interleukin 27 subunit alpha; Interleukin 27; 28-KD subunit; Interleukin 30; Interleukin-27 subunit alpha; p28. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7455