Learn More
Biomatik Corporation Human Lipolysaccharide Binding Protein, Lbp Elisa Kit
Supplier:  Biomatik Corporation EKC34445
Encompass_Preferred
Human Lipolysaccharide Binding Protein, Lbp Elisa Kit for detecting Lipopolysaccharide Binding Protein in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ug/ml. Sensitivity: 0.156ug/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P18428; LBP. Target Synonyms: BPI fold containing family D; member 2; Bpifd2; LBP; LBP_HUMAN; LBP1; Lipopolysaccharide binding protein; Lipopolysaccharide-binding protein; LPS binding protein; Ly88; MGC22233; OTTHUMP00000030965; RP23-407H16.4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7473