Learn More
Biomatik Corporation Human Lysozyme, Lzm Elisa Kit
Supplier:  Biomatik Corporation EKC34508
Encompass_Preferred
Human Lysozyme, Lzm Elisa Kit for detecting Lysozyme, Lzm in serum, plasma, tissue homogenates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P61626; LYZ. Target Synonyms: 1 4 beta N acetylmuramidase C; 1; 4-beta-N-acetylmuramidase C; EC 3.2.1.17; LYSC_HUMAN; Lysosyme; Lysozyme (renal amyloidosis); Lysozyme C; Lysozyme C precursor; LYZ; LZM; Renal amyloidosis. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7475