Learn More
Biomatik Corporation Human Motilin, Mtl Elisa Kit
Supplier:  Biomatik Corporation EKC34649
Encompass_Preferred
Human Motilin, Mtl Elisa Kit for detecting Motilin, Mtl in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.68pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7483