Learn More
Biomatik Corporation Human Nad-Dependent Deacetylase Sirtuin-6 (Sirt6/Sir2L6) Elisa Kit
Supplier:  Biomatik Corporation EKC34731
Encompass_Preferred
Human Nad-Dependent Deacetylase Sirtuin-6 (Sirt6/Sir2L6) Elisa Kit for detecting Sirtuin (Silent Mating Type Information Regulation 2 Homolog) 6 (S. Cerevisiae) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8N6T7; SIRT6. Target Synonyms: 2810449N18Rik; AI043036; Mono ADP ribosyltransferase sirtuin 6; NAD-dependent protein deacetylase sirtuin-6; Regulatory protein SIR2 homolog 6; Regulatory protein SIR2 homolog; SIR2 like 6; SIR2 like protein 6; Sir2 related protein type 6; SIR2-like protein 6; SIR2L6; SIR6_HUMAN; SIRT 6; SIRT6; Sirtuin (silent mating type information regulation 2 homolog) 6 (S. cerevisiae); Sirtuin 6; Sirtuin type 6; Sirtuin6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7488