Learn More
Biomatik Corporation Human Orexin A Elisa Kit
Supplier:  Biomatik Corporation EKC34886
Encompass_Preferred
Human Orexin A Elisa Kit for detecting Orexin A in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O43612; HCRT. Target Synonyms: Hcrt; Hcrt1; Hcrt2; Hypocretin; Hypocretin neuropeptide precursor; Hypocretin-1; Hypocretin-2; OREX_HUMAN; Orexin; Orexin precursor; Orexin-B; OX; PPOX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7499