Learn More
Biomatik Corporation Human Periostin/Osteoblast Specific Factor 2 (Postn) Elisa Kit
Supplier:  Biomatik Corporation EKC34958
Encompass_Preferred
Human Periostin/Osteoblast Specific Factor 2 (Postn) Elisa Kit for detecting Periostin/Osteoblast Specific Factor 2 (Postn) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 1.56ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q15063; POSTN. Target Synonyms: Fasciclin-I like; MGC119510; MGC119511; OSF 2; OSF-2; OSF2; Osteoblast specific factor 2 (fasciclin I like); Osteoblast specific factor 2; Osteoblast specific factor; Osteoblast-specific factor 2; PDLPOSTN; Periodontal ligament specific periostin; Periostin; Periostin isoform thy2; Periostin isoform thy4; Periostin isoform thy6; Periostin isoform thy8; Periostin osteoblast specific factor; PN; POSTN; POSTN_HUMAN; RP11 412K4.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7511