Learn More
Biomatik Corporation Human Plasminogen Activator Inhibitor 1, Pai-1 Elisa Kit
Supplier:  Biomatik Corporation EKC35039
Encompass_Preferred
Human Plasminogen Activator Inhibitor 1, Pai-1 Elisa Kit for detecting Serpin Peptidase Inhibitor, Clade E (Nexin, Plasminogen Activator Inhibitor Type 1), Member 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.125-200ng/ml. Sensitivity: 2.201ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05121; SERPINE1. Target Synonyms: Clade E; Endothelial plasminogen activator inhibitor; Nexin; Nexin plasminogen activator inhibitor type 1; PAI 1; PAI; PAI-1; PAI1_HUMAN; PLANH1; Plasminogen activator inhibitor 1; Plasminogen activator inhibitor type 1; Serine (or cysteine) proteinase inhibitor; Serine (or cysteine) proteinase inhibitor clade E (nexin plasminogen activator inhibitor type 1) member 1; Serine proteinase inhibitor clade E member 1; serpin; Serpin E1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7519